2,4-Difluoro-3-methylaniline 96%

Supplier: Ambeed
Danger

A235118-100MG A235118-1G A235118-250MG A235118-5G
76987-492EA 24.41 USD
76987-492 76987-494 76987-496 77683-696
2,4-Difluoro-3-methylaniline 96%
2,4-Difluoro-3-methylaniline

Precaución: For research use only.

Formula: C₇H₇F₂N
MW: 143.134 g/mol
MDL Number: MFCD09261073
CAS Number: 76350-70-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR