3-Chloro-6-fluoropyridazine 95%

Supplier: Ambeed
Warning

A234878-100MG A234878-1G
76987-482EA 113 USD
76987-482 77550-512
3-Chloro-6-fluoropyridazine 95%
3-Chloro-6-fluoropyridazine

Precaución: for Research Use Only

Formula: C₄H₂ClFN₂
MDL Number: MFCD12547767
CAS Number: 33097-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR