2,6-Dichloro-4-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A234687-100MG A234687-1G
76987-476EA 60.48 USD
76987-476 76987-478
2,6-Dichloro-4-methoxybenzaldehyde 95%
2,6-Dichloro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
MDL Number: MFCD12024958
CAS Number: 82772-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR