2-Amino-4,6-difluorobenzothiazole 98%

Supplier: Ambeed
Warning

A234531-1G A234531-250MG A234531-5G A234531-25G
77000-266EA 15 USD
77000-266 77000-268 77000-270 77881-609
2-Amino-4,6-difluorobenzothiazole 98%
2-Amino-4,6-difluorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂S
MDL Number: MFCD00276099
CAS Number: 119256-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR