4-Methoxy-2,6-dimethylpyrimidine 97%

Supplier: Ambeed
Warning

A234400-100MG A234400-1G A234400-250MG
77013-626EA 64 USD
77013-626 77013-628 77013-630
4-Methoxy-2,6-dimethylpyrimidine 97%
4-Methoxy-2,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MDL Number: MFCD18377698
CAS Number: 14001-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR