Methyl-3,5-dinitrobenzoate 98%

Supplier: Ambeed
Warning

A234369-100G A234369-25G A234369-500G A234369-5G
77000-258EA 86 USD
77000-258 77000-260 77000-262 77000-264
Methyl-3,5-dinitrobenzoate 98%
Methyl-3,5-dinitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆N₂O₆
MDL Number: MFCD00017016
CAS Number: 2702-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR