Methyl 2-((tert-butoxycarbonyl)amino)isonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2,2-Dimethylpropionylamino)isonicotinic acid methyl ester

A234239-100MG A234239-250MG A234239-5G
77031-134EA 18 USD
77031-134 77031-136 77031-138
Methyl 2-((tert-butoxycarbonyl)amino)isonicotinate 97%
Methyl 2-((tert-butoxycarbonyl)amino)isonicotinate

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₄
MDL Number: MFCD11504796
CAS Number: 639091-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR