6-(Chloromethyl)uracil 98%

Supplier: Ambeed
Warning

Synonyms: 6-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione

A234226-10G A234226-250MG A234226-5G A234226-1G A234226-25G
77013-620EA 27 USD
77013-620 77013-622 77013-624 77890-357 77890-358
6-(Chloromethyl)uracil 98%
6-(Chloromethyl)uracil

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
MDL Number: MFCD00010095
CAS Number: 18592-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR