Methyl-5-amino-2-chlorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-amino-2-chlorobenzoate

A234174-100G A234174-10G A234174-1G A234174-5G
77031-126EA 368.14 USD
77031-126 77031-128 77031-130 77031-132
Methyl-5-amino-2-chlorobenzoate 97%
Methyl 5-amino-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.6100 g/mol
MDL Number: MFCD07800315
CAS Number: 42122-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR