4-Bromo-5-chloro-2-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A234004-1G A234004-250MG A234004-25G A234004-5G
77013-612EA 55.17 USD
77013-612 77013-614 77013-616 77013-618
4-Bromo-5-chloro-2-fluorobenzoic acid 98%
4-Bromo-5-chloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
MDL Number: MFCD19686184
CAS Number: 1349708-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR