6-tert-Butyl-3-ethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate, N-BOC protected, Ethyl 6-Boc-2-amino-4,7-dihydro-5H-thieno[2,3-c]pyridine-3-carboxylate

A233959-1G A233959-250MG A233959-5G
77031-122EA 20.16 USD
77031-122 77031-124 77667-934
6-tert-Butyl-3-ethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate 97%
6-tert-Butyl-3-ethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₄S
MW: 326.42 g/mol
MDL Number: MFCD05664039
CAS Number: 193537-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR