5-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one 97%

Supplier: Ambeed
Warning

A233915-100MG A233915-1G A233915-250MG A233915-5G
77013-598EA 32.89 USD
77013-598 77013-600 77013-602 77013-604
5-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one 97%
5-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.05 g/mol
MDL Number: MFCD00504782
CAS Number: 84712-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR