6-Bromo-3-fluoro-o-xylene 95%

Supplier: Ambeed
Warning

A233791-100MG A233791-1G A233791-250MG A233791-5G
76987-460EA 5 USD
76987-460 76987-462 76987-464 76987-466
6-Bromo-3-fluoro-o-xylene 95%
6-Bromo-3-fluoro-o-xylene

Precaución: for Research Use Only

Formula: C₈H₈BrF
MDL Number: MFCD03789115
CAS Number: 52548-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR