6-Fluoro-3-methyl-1H-indazole 95%

Supplier: Ambeed
Warning

A233741-100MG A233741-1G A233741-250MG
77031-116EA 67.9 USD
77031-116 77031-118 77031-120
6-Fluoro-3-methyl-1H-indazole 95%
6-Fluoro-3-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇FN₂
MW: 150.15 g/mol
MDL Number: MFCD08692363
CAS Number: 159305-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR