Chloroquine diphosphate salt 98%

Supplier: Ambeed
Warning

Synonyms: N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate

A233637-100G A233637-100MG A233637-10G A233637-25G A233637-500G A233637-5G A233637-1000G
77013-580EA 39.26 USD
77013-580 77013-582 77013-584 77013-586 77013-588 77013-590 77683-683
Chloroquine diphosphate salt 98%
Chloroquine diphosphate salt

Precaución: For research use only.

Formula: C₁₈H₂₆ClN₃
MW: 515.86 g/mol
MDL Number: MFCD00069852
CAS Number: 50-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR