2-((2-Hydroxyethyl)amino)acetic acid 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-(2-Hydroxyethyl)-Gly-OH

A233372-100MG A233372-250MG
77013-568EA 47.75 USD
77013-568 77683-682
2-((2-Hydroxyethyl)amino)acetic acid 98%, Ambeed.Inc
2-((2-Hydroxyethyl)amino)acetic acid

Precaución: For research use only.

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00082194
CAS Number: 5835-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR