6-Methylisoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A233353-100MG A233353-1G A233353-250MG A233353-5G
77013-560EA 27 USD
77013-560 77013-562 77013-564 77013-566
6-Methylisoquinolin-1(2H)-one 95%
6-Methylisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD11559024
CAS Number: 131002-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR