Methyl imidazo[1,2-a]pyridine-7-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A233244-10G
A233244-1G
A233244-250MG
A233244-5G
77031-104EA
211.12
USD
77031-104
77031-106
77031-108
77031-110
Methyl imidazo[1,2-a]pyridine-7-carboxylate 95%
Methyl imidazo[1,2-a]pyridine-7-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂ MW: 176.18 g/mol |
MDL Number:
MFCD09910350 CAS Number: 86718-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...