1-Chloro-4-(methoxymethyl)benzene 95%

Supplier: Ambeed
Warning

A233088-1G A233088-250MG A233088-5G
77013-546EA 70.02 USD
77013-546 77013-548 77013-550
1-Chloro-4-(methoxymethyl)benzene 95%
1-Chloro-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.609 g/mol
MDL Number: MFCD22054452
CAS Number: 1195-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR