Methyl 4-amino-1-methyl-1H-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

A232991-100MG A232991-1G A232991-250MG A232991-5G
77013-540EA 32.89 USD
77013-540 77013-542 77013-544 77612-280
Methyl 4-amino-1-methyl-1H-pyrazole-3-carboxylate 95%
Methyl 4-amino-1-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.155 g/mol
MDL Number: MFCD04969986
CAS Number: 637336-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR