3-Amino-5-cyclopropylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Cyclopropylpyridin-3-amine

A232756-1G A232756-250MG
76987-448EA 300.24 USD
76987-448 76987-450
3-Amino-5-cyclopropylpyridine 95%
3-Amino-5-cyclopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
MW: 134.178 g/mol
MDL Number: MFCD19543987
CAS Number: 1314353-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR