2-(4-Pentylphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A232734-10G A232734-25G A232734-5G
77000-238EA 56 USD
77000-238 77000-240 77000-242
2-(4-Pentylphenyl)acetic acid 98%
2-(4-Pentylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
MDL Number: MFCD00605331
CAS Number: 14377-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR