Chroman-7-carboxylic acid 95%

Supplier: Ambeed
Warning

A232324-100MG A232324-250MG
76987-430EA 81 USD
76987-430 76987-432
Chroman-7-carboxylic acid 95%
Chroman-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
MDL Number: MFCD14706361
CAS Number: 527681-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR