Ethyl 4,5-dimethyloxazole-2-carboxylate 96%

Supplier: Ambeed
Warning

A232181-100MG A232181-1G A232181-250MG
77013-524EA 121 USD
77013-524 77013-526 77013-528
Ethyl 4,5-dimethyloxazole-2-carboxylate 96%
Ethyl 4,5-dimethyloxazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MDL Number: MFCD22689290
CAS Number: 33123-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR