Methyl 5-bromo-2,4-dimethylbenzoate 95%

Supplier: Ambeed
Warning

A232176-100MG A232176-1G A232176-250MG A232176-50MG
76987-418EA 100 USD
76987-418 76987-420 76987-422 76987-424
Methyl 5-bromo-2,4-dimethylbenzoate 95%
Methyl 5-bromo-2,4-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD11112985
CAS Number: 152849-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR