Dimethyl bicyclo[2.2.2]octane-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

A232111-1G A232111-5G
77031-076EA 22.28 USD
77031-076 77031-078
Dimethyl bicyclo[2.2.2]octane-1,4-dicarboxylate 95%
Dimethyl bicyclo[2.2.2]octane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₄
MW: 226.269 g/mol
MDL Number: MFCD01312167
CAS Number: 1459-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR