2-Chloro-4-methylpyrimidine 98%

Supplier: Ambeed
Warning

A231936-100G A231936-10G A231936-1G A231936-25G A231936-500G A231936-5G
77000-226EA 125.19 USD
77000-226 77000-228 77000-230 77000-232 77000-234 77000-236
2-Chloro-4-methylpyrimidine 98%
2-Chloro-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MDL Number: MFCD00054434
CAS Number: 13036-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR