2-Fluoro-5-methoxyphenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Carboxymethyl)-4-fluoroanisole, (2-Fluoro-5-methoxyphenyl)acetic acid, 2-(2-Fluoro-5-methoxyphenyl)acetic acid

A231934-1G A231934-5G
77031-072EA 42.44 USD
77031-072 77031-074
2-Fluoro-5-methoxyphenylacetic acid 97%
2-Fluoro-5-methoxyphenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
MDL Number: MFCD13185602
CAS Number: 798563-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR