3,4-Dichloro-1,2,5-thiadiazole 97%

Supplier: Ambeed
Warning

A231893-10G A231893-25G A231893-5G
77000-224EA 10.61 USD
77000-224 77089-460 77672-064
3,4-Dichloro-1,2,5-thiadiazole 97%
3,4-Dichloro-1,2,5-thiadiazole

Precaución: for Research Use Only

Formula: C₂Cl₂N₂S
MW: 155.01 g/mol
MDL Number: MFCD00010072
CAS Number: 5728-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR