1-Fluoro-3-methoxy-5-methyl-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A231871-100MG A231871-250MG
77013-514EA 120.95 USD
77013-514 77013-516
1-Fluoro-3-methoxy-5-methyl-2-nitrobenzene 98%
1-Fluoro-3-methoxy-5-methyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
MW: 185.15 g/mol
MDL Number: MFCD26743724
CAS Number: 1631962-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR