7-Chlorobenzo[d]thiazole-2(3H)-thione 97%

Supplier: Ambeed
Warning

A231836-100MG A231836-1G A231836-250MG
76987-412EA 5.31 USD
76987-412 76987-414 76987-416
7-Chlorobenzo[d]thiazole-2(3H)-thione 97%
7-Chlorobenzo[d]thiazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₇H₄ClNS₂
MW: 201.7 g/mol
MDL Number: MFCD11656576
CAS Number: 1849-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR