2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Glycinol, N-Fmoc-ethanolamine, 2-(Fmoc-amino)-1-ethanol

A231712-10G A231712-1G A231712-25G A231712-5G A231712-100G
77000-212EA 11 USD
77000-212 77000-214 77000-216 77000-218 77672-062
2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol 98%
2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol

Precaución: for Research Use Only

Formula: C₁₇H₁₇NO₃
MW: 283.33 g/mol
MDL Number: MFCD00235927
CAS Number: 105496-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR