2-Fluoro-4-formylbenzoic acid 98%

Supplier: Ambeed
Warning

A231671-100MG A231671-1G A231671-250MG A231671-5G
77013-506EA 33 USD
77013-506 77013-508 77013-510 77013-512
2-Fluoro-4-formylbenzoic acid 98%
2-Fluoro-4-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅FO₃
MDL Number: MFCD12026481
CAS Number: 604000-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR