2-Amino-5-bromo-4-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-carboxy-5-fluoroaniline, 5-Bromo-4-fluoroanthranilic acid

A231639-100G A231639-10G A231639-1G A231639-250MG A231639-25G A231639-500G A231639-5G
77031-054EA 167 USD
77031-054 77031-056 77031-058 77031-060 77031-062 77031-064 77031-066
2-Amino-5-bromo-4-fluorobenzoic acid 97%
2-Amino-5-bromo-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD06409112
CAS Number: 143945-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR