2-Bromo-3-iodo-5-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A231600-100MG A231600-1G A231600-250MG A231600-5G
77031-046EA 5 USD
77031-046 77031-048 77031-050 77031-052
2-Bromo-3-iodo-5-(trifluoromethyl)pyridine 95%
2-Bromo-3-iodo-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂BrF₃IN
MW: 351.9 g/mol
MDL Number: MFCD14702319
CAS Number: 1214323-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR