Methyl-3-aminocrotonate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Amino-2-butenoate, 3-Aminocrotonic acid Methyl Ester, Methyl 3-aminobut-2-enoate, 3-Amino-2-butenoic acid Methyl Ester, Methyl 3-Aminocrotonate

A231375-100G A231375-500G A231375-25G
77000-208EA 14.86 USD
77000-208 77000-210 77089-414
Methyl-3-aminocrotonate 95%
Methyl 3-aminocrotonate

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD00008072
CAS Number: 14205-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR