2,4-Dichloro-5-methylpyrimidine 98%

Supplier: Ambeed
Danger

A231363-10G A231363-25G A231363-5G A231363-100G
76987-396EA 9.55 USD
76987-396 76987-398 76987-400 77612-256
2,4-Dichloro-5-methylpyrimidine 98%
2,4-Dichloro-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
MDL Number: MFCD00023197
CAS Number: 1780-31-0
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR