1-Ethyl-3-hydroxypiperidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Ethylpiperidin-3-ol, 1-Ethyl-3-piperidinol

A231304-100G A231304-25G A231304-5G
77000-202EA 165.51 USD
77000-202 77000-204 77000-206
1-Ethyl-3-hydroxypiperidine 98%
1-Ethyl-3-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MW: 129.20 g/mol
MDL Number: MFCD00006508
CAS Number: 13444-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR