7-Bromo-2-methylquinoline 97%

Supplier: Ambeed
Warning

A231279-100MG A231279-1G A231279-250MG A231279-5G
77031-038EA 17 USD
77031-038 77031-040 77031-042 77031-044
7-Bromo-2-methylquinoline 97%
7-Bromo-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD09787847
CAS Number: 4965-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR