5-Bromo-2-fluoro-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-fluoro-3-pyridylboronic, 5-Bromo-2-fluoro-3-pyridinylboronic acid, 5-Bromo-2-fluoropyridine-3-boronic acid

A231277-1G A231277-25G A231277-5G
76987-388EA 9 USD
76987-388 76987-390 76987-392
5-Bromo-2-fluoro-3-pyridineboronic acid 98%
5-Bromo-2-fluoro-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BBrFNO₂
MDL Number: MFCD04039317
CAS Number: 501435-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR