2-(N-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethyl)-2-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetamido)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-PNA-T-OH

A231178-100MG A231178-250MG A231178-5G A231178-1G
76987-384EA 37 USD
76987-384 76987-386 77568-236 77612-250
2-(N-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethyl)-2-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetamido)acetic acid 95%
2-(N-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethyl)-2-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetamido)acetic acid

Precaución: For research use only.

Formula: C₂₆H₂₆N₄O₇
MDL Number: MFCD01940758
CAS Number: 169396-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR