5-Bromo-4-chloro-2,6-dimethylpyrimidine 97%

Supplier: Ambeed
Warning

A231133-100MG A231133-250MG A231133-5G A231133-1G
76987-378EA 76 USD
76987-378 76987-380 76987-382 77890-347
5-Bromo-4-chloro-2,6-dimethylpyrimidine 97%
5-Bromo-4-chloro-2,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₆BrClN₂
MDL Number: MFCD13705450
CAS Number: 69696-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR