4-Chloro-5-nitro-1H-indazole 97+%

Supplier: Ambeed
Warning

A231103-1G A231103-250MG
76987-374EA 172.93 USD
76987-374 76987-376
4-Chloro-5-nitro-1H-indazole 97+%
4-Chloro-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MW: 197.57 g/mol
MDL Number: MFCD11007841
CAS Number: 907958-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR