(S)-3-Amino-3-(3-bromophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A231093-1G A231093-250MG A231093-5G
77000-196EA 107.16 USD
77000-196 77000-198 77000-200
(S)-3-Amino-3-(3-bromophenyl)propanoic acid 98%
(S)-3-Amino-3-(3-bromophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.08 g/mol
MDL Number: MFCD04113686
CAS Number: 275826-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR