Piperidin-3-one hydrochloride 98%

Supplier: Ambeed
Warning

A231091-100G A231091-1G A231091-5G A231091-25G
77013-500EA 152 USD
77013-500 77013-502 77013-504 77550-494
Piperidin-3-one hydrochloride 98%
Piperidin-3-one hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO
MDL Number: MFCD06796739
CAS Number: 61644-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR