4-(2-Propynyloxy)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Aminophenyl Propargyl ether, 4-(Propargyloxy)aniline

A230770-1G A230770-5G A230770-100MG
77013-496EA 111.4 USD
77013-496 77568-230 77683-671
4-(2-Propynyloxy)aniline 98%
4-(2-Propynyloxy)aniline

Precaución: For research use only.

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD12823190
CAS Number: 26557-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR