4-Bromo-2-(2,2,2-trifluoroethoxy)pyridine 95%

Supplier: Ambeed
Warning

A230596-250MG A230596-1G
76987-346EA 60 USD
76987-346 77890-344
4-Bromo-2-(2,2,2-trifluoroethoxy)pyridine 95%
4-Bromo-2-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
MDL Number: MFCD13190652
CAS Number: 161952-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR