2-(4-(3-Chloropropyl)piperazin-1-yl)ethanol dihydrochloride 95%

Supplier: Ambeed
Warning

A230544-1G A230544-250MG A230544-5G
76987-340EA 20.16 USD
76987-340 76987-342 76987-344
2-(4-(3-Chloropropyl)piperazin-1-yl)ethanol dihydrochloride 95%
2-(4-(3-Chloropropyl)piperazin-1-yl)ethanol dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₁Cl₃N₂O
MW: 279.64 g/mol
MDL Number: MFCD09752083
CAS Number: 3445-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR