4-Amino-5-methylpyridin-2-ol 95%

Supplier: Ambeed
Warning

A230512-1G A230512-250MG A230512-5G
76987-334EA 66.84 USD
76987-334 76987-336 76987-338
4-Amino-5-methylpyridin-2-ol 95%
4-Amino-5-methylpyridin-2-ol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD12546373
CAS Number: 95306-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR