2-Ethoxy-5-methylaniline 97%

Supplier: Ambeed
Warning

A230422-1G A230422-250MG A230422-5G
77000-174EA 24.01 USD
77000-174 77000-176 77000-178
2-Ethoxy-5-methylaniline 97%
2-Ethoxy-5-methylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD00035775
CAS Number: 6331-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR